Skip to Content
Merck
All Photos(1)

Key Documents

860547P

Avanti

C18 Glucosyl(β) Ceramide (d18:1/18:0)

Avanti Research - A Croda Brand

Synonym(s):

D-glucosyl-β-1,1′-N-stearoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$346.00

$346.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$346.00

About This Item

Empirical Formula (Hill Notation):
C42H81NO8
CAS Number:
Molecular Weight:
728.09
UNSPSC Code:
12352211
NACRES:
NA.25

$346.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 100 mg (860547P-100mg)
pkg of 1 × 25 mg (860547P-25mg)
pkg of 1 × 5 mg (860547P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O

InChI

1S/C42H81NO8/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-38(46)43-35(34-50-42-41(49)40(48)39(47)37(33-44)51-42)36(45)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h29,31,35-37,39-42,44-45,47-49H,3-28,30,32-34H2,1-2H3,(H,43,46)/b31-29+/t35-,36+,37?,39+,40?,41?,42+/m0/s1

InChI key

YMYQEDCYNANIPI-KNSMPFANSA-N

Application

C18 Glucosyl(β) Ceramide (d18:1/18:0) or D-glucosyl-β-1,1′-N-stearoyl-D-erythro-sphingosine has been used:
  • as a lipid standard for its quantification using differential ion mobility spectrometry (DMS) coupled with liquid chromatography in electrospray ionization tandem mass spectrometry (LC-ESI-DMS-MS/MS) in human plasma[1]
  • in the preparation of lipid dispersions[2]
  • used in an assay to monitor the turnover of glucosylceramide in glucocerebrosidase (GBA1) cell free system[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860547P-100mg)
5 mL Amber Glass Screw Cap Vial (860547P-25mg)
5 mL Amber Glass Screw Cap Vial (860547P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong Hoon Won et al.
Neuroscience, 427, 1-15 (2019-11-26)
Neurotransmitter release is mediated by ceramide, which is generated by sphingomyelin hydrolysis. In the present study, we examined whether synaptosomal-associated protein 25 (SNAP-25) is involved in ceramide production and exocytosis. Neutral sphingomyelinase 2 (nSMase2) was partially purified from bovine brain
Differential mobility spectrometry as an orthogonal separation to LC-ESI-MS/MS for quantification of isomeric cerebrosides in plasma and cerebrospinal fluid
Xu H, et al.
Journal of Lipid Research, jlr-D089797 (2018)
Hongbin Xu et al.
Journal of lipid research, 60(1), 200-211 (2018-11-11)
Cerebrosides, including glucosylceramides (GlcCers) and galactosylceramides (GalCers), are important membrane components of animal cells with deficiencies resulting in devastating lysosomal storage disorders. Their quantification is essential for disease diagnosis and a better understanding of disease mechanisms. The simultaneous quantification of
Joseph R Mazzulli et al.
Cell, 146(1), 37-52 (2011-06-28)
Parkinson's disease (PD), an adult neurodegenerative disorder, has been clinically linked to the lysosomal storage disorder Gaucher disease (GD), but the mechanistic connection is not known. Here, we show that functional loss of GD-linked glucocerebrosidase (GCase) in primary cultures or
Victoria Schiffer et al.
PloS one, 15(1), e0227077-e0227077 (2020-01-14)
Gaucher disease is caused by a deficiency in glucocerebrosidase that can result in non-neuronal as well as neuronal symptoms. Common visceral symptoms are an increased organ size, specifically of the spleen, and glucosylceramide as well as glucosylsphingosine substrate accumulations as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service