Skip to Content
Merck
All Photos(1)

Key Documents

860542P

Avanti

Lactosyl(β) Sphingosine (d18:1)

Avanti Research - A Croda Brand 860542P, powder

Synonym(s):

D-lactosyl-β1-1′-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$476.00

$476.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$476.00

About This Item

Empirical Formula (Hill Notation):
C30H57NO12
CAS Number:
Molecular Weight:
623.77
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$476.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860542P-5mg)
pkg of 1 × 50 mg (860542P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860542P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@](N)([H])CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O

InChI

1S/C30H57NO12/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-20(34)19(31)18-40-29-27(39)25(37)28(22(17-33)42-29)43-30-26(38)24(36)23(35)21(16-32)41-30/h14-15,19-30,32-39H,2-13,16-18,31H2,1H3/b15-14+/t19-,20+,21+,22+,23-,24-,25+,26+,27+,28+,29+,30-/m0/s1

InChI key

MQKSCOKUMZMISB-GPWKTZPCSA-N

General description

Lactosyl sphingosine is a glycosylsphingosine (Lyso-GSL), which is found in bovine brain white matter extract.[1]

Application

Lactosyl(β) Sphingosine (d18:1) or D-lactosyl-β1-1′-D-erythro-sphingosine has been used as an internal standard for the quantification of psychosine using tandem mass spectrometry.[2][3] It may be used to screen lipids that rapidly and reversibly alter transepithelial electrical resistance (TER) or tight junction (TJ) permeability in epithelial tissue.[4] It may also be used as a standard to determine the extent of recovery of lyso-neutral glycosphingolipids (lyso-n-GSLs) from the Sephadex column.[5]

Biochem/physiol Actions

Lactosyl sphingosine exhibits better hemolytic activity than lysolecithin.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (860542P-50mg)
5 mL Amber Glass Screw Cap Vial (860542P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhimankrishna Ghosh et al.
Cell systems, 4(5), 516-529 (2017-04-04)
We present a systems strategy that facilitated the development of a molecular signature for glioblastoma (GBM), composed of 33 cell-surface transmembrane proteins. This molecular signature, GBMSig, was developed through the integration of cell-surface proteomics and transcriptomics from patient tumors in
Amy C Gross et al.
JCI insight, 3(16) (2018-08-24)
Osteosarcoma (OS), a malignant tumor of bone, kills through aggressive metastatic spread almost exclusively to the lung. Mechanisms driving this tropism for lung tissue remain unknown, though likely invoke specific interactions between tumor cells and other cells within the lung
Michael S Marshall et al.
PloS one, 13(2), e0193438-e0193438 (2018-02-27)
α-Synuclein aggregation has been linked to Gaucher's disease (GD) and Krabbe's disease (KD), lysosomal conditions affecting glycosphingolipid metabolism. α-Synuclein pathology has been directly attributed to the dysregulation of glycosphingolipids in both conditions, specifically to increased galactosylsphingosine (psychosine) content in the
Richard Mair et al.
Cancer research, 78(18), 5408-5418 (2018-07-29)
13C MRI of hyperpolarized [1-13C]pyruvate metabolism has been used in oncology to detect disease, investigate disease progression, and monitor response to treatment with a view to guiding treatment in individual patients. This technique has translated to the clinic with initial
Preparation of lysohematoside from hematoside of equine erythrocyte and its chemical and hemolytic properties.
T Taketomi et al.
Journal of biochemistry, 68(4), 475-485 (1970-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service