Skip to Content
Merck
All Photos(1)

Key Documents

860466P

Avanti

N-C12-desoxymethylsphingosine

Avanti Research - A Croda Brand 860466P, powder

Synonym(s):

N-lauroyl-1-desoxymethylsphingosine (m17:1/12:0)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$450.00

$450.00


Estimated to ship onApril 18, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$450.00

About This Item

Empirical Formula (Hill Notation):
C29H57NO2
CAS Number:
Molecular Weight:
451.77
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$450.00


Estimated to ship onApril 18, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (860466P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860466P

lipid type

sphingolipids
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)CNC(CCCCCCCCCCC)=O

InChI key

NQTIAYDGVUCKFJ-KSPVXFDASA-N

General description

Desoxymethylsphingosine is an atypical deoxy-sphingoid base.[1]

Application

N-C12-desoxymethylsphingosine or N-lauroyl-1-desoxymethylsphingosine (m17:1/12:0) has been used as a standard in the quantitation of atypical sphingoid bases in biological samples by reverse-phase liquid chromatography coupled to electrospray ionization tandem mass spectrometry (LC-MS/MS).[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860466P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Marshall et al.
PloS one, 13(2), e0193438-e0193438 (2018-02-27)
α-Synuclein aggregation has been linked to Gaucher's disease (GD) and Krabbe's disease (KD), lysosomal conditions affecting glycosphingolipid metabolism. α-Synuclein pathology has been directly attributed to the dysregulation of glycosphingolipids in both conditions, specifically to increased galactosylsphingosine (psychosine) content in the
Michael S Marshall et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 26(3), 874-889 (2018-02-13)
We report a global adeno-associated virus (AAV)9-based gene therapy protocol to deliver therapeutic galactosylceramidase (GALC), a lysosomal enzyme that is deficient in Krabbe's disease. When globally administered via intrathecal, intracranial, and intravenous injections to newborn mice affected with GALC deficiency
Junliang Wan et al.
Journal of agricultural and food chemistry, 67(46), 12953-12961 (2019-10-23)
Most common sphingolipids are comprised of "typical" sphingoid bases (sphinganine, sphingosine, and structurally related compounds) and are produced via the condensation of l-serine with a fatty acyl-CoA by serine palmitoyltransferase. Some organisms, including mammals, also produce "atypical" sphingoid bases that
Sarah T Pruett et al.
Journal of lipid research, 49(8), 1621-1639 (2008-05-24)
"Sphingosin" was first described by J. L. W. Thudichum in 1884 and structurally characterized as 2S,3R,4E-2-aminooctadec-4-ene-1,3-diol in 1947 by Herb Carter, who also proposed the designation of "lipides derived from sphingosine as sphingolipides." This category of amino alcohols is now
Terina N Martinez et al.
Molecular neurodegeneration, 7, 45-45 (2012-09-15)
Dopaminergic (DA) neurons in the ventral midbrain selectively degenerate in Parkinson's disease (PD) in part because their oxidative environment in the substantia nigra (SN) may render them vulnerable to neuroinflammatory stimuli. Chronic inhibition of soluble Tumor Necrosis Factor (TNF) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service