Skip to Content
Merck
All Photos(2)

Key Documents

860450P

Avanti

14:0 PA-d54

Avanti Research - A Croda Brand, powder

Synonym(s):

1,2-Dimyristoyl-d54-sn-glycero-3-phosphate, sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,490.00

$1,490.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,490.00

About This Item

Empirical Formula (Hill Notation):
C31H6O8PNaD54
CAS Number:
Molecular Weight:
669.10
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$1,490.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860450P-10MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(O)([O-])=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])

General description

1,2-dimyristoyl-d54-sn-glycero-3-phosphate (sodium salt) (14:0 PA-d54) is a deuterated phosphatidic acid in which 54 protons of dimyristoyl are replaced by deuterium isotope. Phosphatidic acid (PA) is a membrane phospholipid.[1]

Biochem/physiol Actions

Phosphatidic acid (PA) serves as an intermediate for the production of storage and membrane lipids. It is involved in the regulation of glycerolipid metabolism. PA is also involved in membrane biogenesis. It is also implicated in several cellular processes such as, membrane trafficking, signal transduction, cytoskeletal rearrangement and secretion.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860450P-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Signaling functions of phosphatidic acid
Wang X, et al.
Progress in Lipid Research, 45(3), 250-278 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service