Skip to Content
Merck
All Photos(2)

Key Documents

860362P

Avanti

18:0 PC-d9

Avanti Research - A Croda Brand 860362P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphocholine-N,N,N-trimethyl-d9

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,410.00

$1,410.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$1,410.00

About This Item

Empirical Formula (Hill Notation):
C44H79NO8PD9
CAS Number:
Molecular Weight:
799.20
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$1,410.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860362P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860362P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1/i3D3,4D3,5D3

InChI key

NRJAVPSFFCBXDT-BABXQRPRSA-N

General description

18:0 PC-d9 is a deuterated phosphocholine (PC) in which nine protons of distearoyl are replaced by deuterium.

Application

18:0 PC-d9 has been used as a standard for the quantification of choline and choline metabolites from milk and blood samples using liquid chromatography-tandem mass spectrometry.[1][2]

Packaging

5 mL Amber Glass Screw Cap Vial (860362P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choline and choline metabolite patterns and associations in blood and milk during lactation in dairy cows
Artegoitia VM, et al.
PLoS ONE, 9(8) (2014)
Choline and choline metabolite patterns and associations in blood and milk during lactation in dairy cows
Artegoitia VM, et al.
Testing, 9(8) (2014)
Feeding increasing amounts of ruminally protected choline decreased fatty liver in nonlactating, pregnant Holstein cows in negative energy status
Zenobi MG, et al.
Journal of Dairy Science, 101(7), 5902-5923 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service