Skip to Content
Merck
All Photos(1)

Key Documents

858152L

Avanti

17:0-16:1 PS-d5

Avanti Research - A Croda Brand

Synonym(s):

1-heptadecanoyl-2-palmitoleoyl-sn-glycero(d5)-3-phospho- L-serine (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,230.00

$1,230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C39H68D5NNaO10P
CAS Number:
Molecular Weight:
774.99
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

$1,230.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

solution

packaging

package of 1 × 1 mg

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (858152L-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service