Skip to Content
Merck
All Photos(1)

Key Documents

857511P

Avanti

Iso 15:0 Fatty Acid

Avanti Research - A Croda Brand (857511P), powder

Synonym(s):

13-Methyltetradecanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$203.00
25 MG
$405.00

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$203.00
25 MG
$405.00

About This Item

Empirical Formula (Hill Notation):
C15H30O2
CAS Number:
Molecular Weight:
242.40
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (857511P-10MG)
pkg of 1 × 25 mg (857511P-25MG)

manufacturer/tradename

Avanti Research - A Croda Brand (857511P)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)CCCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-14(2)12-10-8-6-4-3-5-7-9-11-13-15(16)17/h14H,3-13H2,1-2H3,(H,16,17)

InChI key

ZOCYQVNGROEVLU-UHFFFAOYSA-N

General description

Iso 15:0 Fatty Acid, also known as 13-Methyltetradecanoic acid, belongs to the family of branched chain fatty acids (BCFA).[1] It is obtained from soy fermentation products.[2]

Biochem/physiol Actions

Iso 15:0 fatty acid possesses anti-cancer activity in various human cancer cells.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (857511P-10MG and 857511P-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

13-Methyltetradecanoic acid exhibits anti-tumor activity on T-cell lymphomas in vitro and in vivo by down-regulating p-AKT and activating caspase-3
Cai Q, et al.
PLoS ONE, 8(6), 1669-1676 (2013)
A comprehensive study of serum odd-and branched-chain fatty acids in patients with excess weight
Mika A, et al.
Obesity (Silver Spring, Md.), 24(8), 1669-1676 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service