Skip to Content
Merck
All Photos(2)

Key Documents

855576P

Avanti

15:0 Lyso PC

1-pentadecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-pentadecanoyl-sn-glycero-3-phosphocholine; PC(15:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$284.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$284.00

About This Item

Empirical Formula (Hill Notation):
C23H48NO7P
CAS Number:
Molecular Weight:
481.60
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855576P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855576P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCCCCCC)=O

InChI key

RJZVWDTYEWCUAR-JOCHJYFZSA-N

Application

15:0 Lyso PC may be used:
  • as a reference standard to study the effects of metformin on urinary metabolites in diet-induced-obese mice by ultra-performance liquid chromatography coupled to time-of-flight mass spectrometry (UPLC-TOF/MS)[1]
  • as a standard for the quantification of high-density lipoprotein (HDL) lipidome[2]
  • for the preparation of multilamellar vesicles[3]

Packaging

5 mL Amber Glass Screw Cap Vial (855576P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sterols have higher affinity for sphingomyelin than for phosphatidylcholine bilayers even at equal acyl-chain order
Lonnfors M, et al.
Biophysical Journal, 100(11), 2633-2641 (2011)
Effect of metformin on the urinary metabolites of diet-induced-obese mice studied by ultra performance liquid chromatography coupled to time-of-flight mass spectrometry (UPLC-TOF/MS)
Zhu Y, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 925, 110-116 (2013)
Distinct phospholipid and sphingolipid species are linked to altered HDL function in apolipoprotein AI deficiency
Zakiev E, et al.
Journal of Clinical Lipidology, 13(3), 468-480 (2019)
Untargeted lipidomic analysis of plasma from high fat diet-induced obese rats using UHPLC- linear trap quadrupole -Orbitrap MS.
Siddabasave Gowda B Gowda et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2020-01-21)
Luca Narduzzi et al.
Drug testing and analysis, 12(7), 887-899 (2020-04-05)
Growth hormone (GH), an endogenous peptide regulating anabolism and lipolysis in humans, is known to be abused by athletes to improve their performance. Despite the development of two distinct screening methods, few positive cases have been reported by the antidoping

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service