Skip to Content
Merck
All Photos(2)

Key Documents

850704P

Avanti

15:0 PE

1,2-dipentadecanoyl-sn-glycero-3-phosphoethanolamine, powder

Synonym(s):

PE(15:0/15:0); 110885

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$284.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$284.00

About This Item

Empirical Formula (Hill Notation):
C35H70NO8P
CAS Number:
Molecular Weight:
663.91
UNSPSC Code:
51191904
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850704P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850704P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCC)=O

InChI

1S/C35H70NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-34(37)41-31-33(32-43-45(39,40)42-30-29-36)44-35(38)28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32,36H2,1-2H3,(H,39,40)/t33-/m1/s1

InChI key

SKVKIGSFTGVBOX-MGBGTMOVSA-N

General description

Phosphatidylethanolamine (PE) is a glycerophospholipid.[1]

Application

1,2-dipentadecanoyl-sn-glycero-3-phosphoethanolamine (15:0 PE):
  • has been used as an internal standard for BXD plasma samples in lipidomics analysis[2]
  • may be used as a phospholipid internal standard to analyze phospholipid species from a bacterial culture[3]
  • may be used as an internal standard in ultra-pressure liquid chromatography coupled to high-resolution quadrupole time-of-flight mass spectrometry analysis to study blood plasma lipidome profile[4]
  • may be used as an internal standard in 2D mass spectrometric analysis of hepatic lipid classes[5]

Biochem/physiol Actions

Phosphoethanolamine (PE) serves as a precursor in membrane phospholipid biosynthesis.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (850704P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianlin Han et al.
Analytical biochemistry, 330(2), 317-331 (2004-06-19)
Cellular lipids are composed of linear combinations of aliphatic chains covalently attached to backbones and/or head groups that collectively represent the cellular lipidome. Herein we present a novel two-dimensional electrospray ionization mass spectrometric approach to fingerprint most of the major
Mark Politz et al.
Bio-protocol, 3(21) (2013-11-05)
This protocol describes two similar methods for the extraction and methylation of fatty acids from bacterial cultures. The acid derivatization protocol (Lennen et al., 2013; Bligh and Dyer, 1959) results in the extraction and methylation of all fatty acids, both
Radha M Golla et al.
ACS infectious diseases (2020-04-29)
Staphylococcus aureus is notoriously known for its rapid development of resistance to conventional antibiotics. S. aureus can alter its membrane composition to reduce the killing effect of antibiotics and antimicrobial peptides (AMPs). To obtain a more complete picture, this study
S Imhasly et al.
BMC veterinary research, 11, 252-252 (2015-10-09)
The transition period of dairy cows, around parturition and the onset of lactation, involves endocrine and metabolic changes to compensate for an increased energy requirement aggravated by reduced feed intake. Transition cows adjust to the resulting negative energy balance with
Federica Gibellini et al.
IUBMB life, 62(6), 414-428 (2010-05-27)
The glycerophospholipids phosphatidylcholine (PC) and phosphatidylethanolamine (PE) account for greater than 50% of the total phospholipid species in eukaryotic membranes and thus play major roles in the structure and function of those membranes. In most eukaryotic cells, PC and PE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service