Skip to Content
Merck
All Photos(2)

Key Documents

850536O

Avanti

8.8 MAG

Avanti Research - A Croda Brand 850536O

Synonym(s):

1-(8Z-hexadecenoyl)-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$293.00

$293.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$293.00

About This Item

Empirical Formula (Hill Notation):
C19H36O4
CAS Number:
Molecular Weight:
328.49
UNSPSC Code:
12352211
NACRES:
NA.25

$293.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

oil

mol wt

328.49 g/mol

packaging

pkg of 10 × 0.1 g (with screw cap (850536O-1g))
pkg of 1 × 25 mg (with screw cap (850536O-25mg))

manufacturer/tradename

Avanti Research - A Croda Brand 850536O

technique(s)

protein sequencing: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

OCC(O)COC(CCCCCC/C=C\CCCCCCC)=O

InChI

1S/C19H36O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h8-9,18,20-21H,2-7,10-17H2,1H3/b9-8-

InChI key

CRXZGWVXXGFOLB-HJWRWDBZSA-N

General description

Monoacylglycerols (MAGs) are glycerol esters, that can be produced by the glycerolysis of triglycerides and alkyl esters.[1] MAGs are polar lipids and amphiphilic in nature.[2]
This new Lipid detergent may be used for crystallizing membrane proteins by using lipidic mesophases. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that it has been used to solve X-ray crystallographic structures of prokaryotic and eukaryotic proteins, proteins that are monomeric, homo- and hetero-multimeric, chromophore-containing and chromophore-free, and alpha-helical and beta-barrel proteins.

Application

8.8 MAG may be used as a host lipid to grow MdfA (multidrug transporter protein)–YN1074 (Fab fragment) complex grown using the lipidic cubic phase (LCP) crystallization method.[3]

Biochem/physiol Actions

Monoacylglycerols are used in lotions, ointments and deodorants. It serves as a lubricant, antistatic and plastifying agent, hence it is very useful in the plastic industry.[1]

Packaging

5 mL PTFE Vial with Screw Cap (850536O-1g)
5 mL PTFE Vial with Screw Cap (850536O-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An overview of the development and applications of nanoscale materials in the food industry
Nanotechnology in the Food, Beverage and Nutraceutical Industries (2012)
Kumar Nagarathinam et al.
Acta crystallographica. Section F, Structural biology communications, 73(Pt 7), 423-430 (2017-07-12)
The active efflux of antibiotics by multidrug-resistance (MDR) transporters is a major pathway of drug resistance and complicates the clinical treatment of bacterial infections. MdfA is a member of the major facilitator superfamily (MFS) from Escherichia coli and provides resistance
Synthesis and characterization of monoacylglycerols through glycerolysis of ethyl esters derived from linseed oil by green processes
Hobuss CB, et al.
Royal Society of Chemistry Advances, 10, 2327-2336 (2020)
Niklaus Johner et al.
Journal of the American Chemical Society, 136(8), 3271-3284 (2014-02-06)
The recent advances in the in meso crystallization technique for the structural characterization of G-protein coupled receptor (GPCR) proteins have established the usefulness of the lipidic-cubic phases (LCPs) in the field of crystallography of membrane proteins. It is surprising that
Martin Caffrey et al.
Journal of visualized experiments : JoVE, (45) (2010-11-30)
A detailed protocol for crystallizing membrane proteins by using lipidic mesophases is described. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service