Skip to Content
Merck
All Photos(2)

Key Documents

850534O

Avanti

7.9 MAG

Avanti Research - A Croda Brand 850534O

Synonym(s):

1-(7Z-hexadecenoyl)-rac-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$856.00
1 G
$3,260.00

$856.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$856.00
1 G
$3,260.00

About This Item

Empirical Formula (Hill Notation):
C19H36O4
CAS Number:
Molecular Weight:
328.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$856.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

oil

mol wt

328.49 g/mol

packaging

pkg of 10 × 0.1 g (with screw cap (850534O-1g))
pkg of 2 × 100 mg (with screw cap (850534O-200mg))
pkg of 1 × 25 mg (with screw cap (850534O-25mg))

manufacturer/tradename

Avanti Research - A Croda Brand 850534O

technique(s)

protein sequencing: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

OCC(O)COC(CCCCC/C=C\CCCCCCCC)=O

InChI

1S/C19H36O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h9-10,18,20-21H,2-8,11-17H2,1H3/b10-9-

InChI key

OWGMAJRKMYGYBQ-KTKRTIGZSA-N

General description

This new Lipid detergent may be used for crystallizing membrane proteins by using lipidic mesophases. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that it has been used to solve X-ray crystallographic structures of prokaryotic and eukaryotic proteins, proteins that are monomeric, homo- and hetero-multimeric, chromophore-containing and chromophore-free, and alpha-helical and beta-barrel proteins.

Application

7.9 MAG is suitable for use as a hosting lipid for titration to avoid lamellar crystal phase (Lc) formation during sample injection[1] It is also suitable for use in mesocrystallization.[2]

Biochem/physiol Actions

7.9 MAG is a non-monoolein lipid.[2] It can support crystallization of bacteriorhodopsin, an integral membrane protein.[3]

Packaging

5 mL PTFE Vial with Screw Cap (850534O-1g)
5 mL PTFE Vial with Screw Cap (850534O-200mg)
5 mL PTFE Vial with Screw Cap (850534O-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane protein crystallization in lipidic mesophases with tailored bilayers
Misquitta LV, et al.
Structure, 12(12), 2113-2124 (2004)
In Meso Crystallization of the Integral Membrane Glycerol 3-Phosphate Acyltransferase with Substrates
Li Z, et al.
Crystal Growth & Design, 18(4), 2243-2258 (2018)
Preparation and delivery of protein microcrystals in lipidic cubic phase for serial femtosecond crystallography
Ishchenko A, et al.
Journal of Visualized Experiments, (115), e54463-e54463 (2016)
Niklaus Johner et al.
Journal of the American Chemical Society, 136(8), 3271-3284 (2014-02-06)
The recent advances in the in meso crystallization technique for the structural characterization of G-protein coupled receptor (GPCR) proteins have established the usefulness of the lipidic-cubic phases (LCPs) in the field of crystallography of membrane proteins. It is surprising that
Martin Caffrey et al.
Journal of visualized experiments : JoVE, (45) (2010-11-30)
A detailed protocol for crystallizing membrane proteins by using lipidic mesophases is described. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service