Skip to Content
Merck
All Photos(2)

Key Documents

850468C

Avanti

18:0-18:2 PC

1-stearoyl-2-linoleoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphocholine; PC(18:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$375.00
200 MG
$1,220.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$375.00
200 MG
$1,220.00

About This Item

Empirical Formula (Hill Notation):
C44H84NO8P
CAS Number:
Molecular Weight:
786.11
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850468C-25mg)
pkg of 2 × 4 mL (850468C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850468C

concentration

10 mg/mL (850468C-25mg)
25 mg/mL (850468C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C44H84NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h15,17,21,23,42H,6-14,16,18-20,22,24-41H2,1-5H3/b17-15-,23-21-/t42-/m1/s1

InChI key

FORFDCPQKJHEBF-VPUSDGANSA-N

General description

18:0-18:2 PC is a non-fragmented oxidized phosphotidylcholine.[1] It is a component of low density lipoprotein (LDL), which is a polyunsaturated fatty acid.[2]

Application

18:0-18:2 PC may be used to study the effect of phospholipids on lubrication of damaged mesothelium of lungs.[3] It has been used for the preparation of liposomes.[4]

Packaging

5 mL Clear Glass Sealed Ampule (850468C-200mg)
5 mL Clear Glass Sealed Ampule (850468C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidized phosphatidylcholines are produced in renal ischemia reperfusion injury
Solati Z, et al.
PLoS ONE, 13(4), e0195172-e0195172 (2018)
Lubricating recovery of damaged pleural mesothelium: effect of time and of phosphatidylcholines
Bodega F, et al.
Respiratory Physiology & Neurobiology, 203(2), 116-120 (2014)
Differential ligand binding specificities of the pulmonary collectins are determined by the conformational freedom of a surface loop
Rynkiewicz M J, et al.
Biochemistry, 56(31), 4095-4105 (2017)
Ginger L Milne et al.
Journal of lipid research, 46(2), 307-319 (2004-11-18)
Phospholipids reside in the surface layer of LDLs and constitute approximately 20-25% of the particle by weight. We report a study of the primary products generated from the most abundant molecular species of phosphatidylcholines present in LDL during in vitro
Wenbo Cao et al.
Analytical chemistry, 90(17), 10286-10292 (2018-08-11)
The presence of carbon-carbon double bonds (C═Cs) in unsaturated phospholipids is closely related to lipid conformations and physiochemical activities. Previously, we have demonstrated that epoxidation reaction facilitated by low-temperature plasma (LTP) enabled the structural analysis of unsaturated fatty acids (FAs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service