Skip to Content
Merck
All Photos(2)

Key Documents

850465P

Avanti

18:0-16:0 PC

1-stearoyl-2-palmitoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-octadecanoyl-2-hexadecanoyl-sn-glycero-3-phosphocholine; SPPC; PC(18:0/16:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$189.00
200 MG
$518.00
500 MG
$946.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$189.00
200 MG
$518.00
500 MG
$946.00

About This Item

Empirical Formula (Hill Notation):
C42H84NO8P
CAS Number:
Molecular Weight:
762.09
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850465P-200mg)
pkg of 1 × 25 mg (850465P-25mg)
pkg of 1 × 500 mg (850465P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850465P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H84NO8P/c1-6-8-10-12-14-16-18-20-21-23-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43(3,4)5)51-42(45)35-33-31-29-27-25-22-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3/t40-/m1/s1

InChI key

BYSIMVBIJVBVPA-RRHRGVEJSA-N

General description

Phosphatidylcholine (PC), a phospholipid carries a positively charged quaternary amine and a negatively charged phosphate in its headgroup.[1]

Application

18:0-16:0 PC (1-stearoyl-2-palmitoyl-sn-glycero-3-phosphocholine) may be used:
  • in bilayer membranes to study the thermodynamic properties of phase transitions[2]
  • as a carrier in unilamellar vesicles to evaluate the fragmentation of hydroperoxy endoperoxide 13-HP-Endo-PC in a model membrane[3]
  • as a lipid standard in liquid chromatography−mass spectrometry analysis[4]

Biochem/physiol Actions

Phosphatidylcholine (PC) can serve as a surfactant in the mucus.[5]

Packaging

20 mL Clear Glass Screw Cap Vial (850465P-500mg)
5 mL Amber Glass Screw Cap Vial (850465P-200mg)
5 mL Amber Glass Screw Cap Vial (850465P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Lipid-Protein Interactions in Membranes
The Membranes of Cells, 291-334 (2016)
Xiaodong Gu et al.
Chemical research in toxicology, 24(7), 1080-1093 (2011-05-17)
Biologically active phospholipids that incorporate an oxidatively truncated acyl chain terminated by a γ-hydroxyalkenal are generated in vivo. The γ-hydroxyalkenal moiety protrudes from lipid bilayers like whiskers that serve as ligands for the scavenger receptor CD36, fostering endocytosis, e.g., of
Masaki Goto et al.
Biochimica et biophysica acta, 1778(4), 1067-1078 (2008-01-15)
The bilayer phase transitions of palmitoylstearoyl-phosphatidylcholine (PSPC), diheptadecanoyl-PC (C17PC) and stearoylpalmitoyl-PC (SPPC) which have the same total carbon numbers in the two acyl chains were observed by differential scanning calorimetry and high-pressure optical method. As the temperature increased, these bilayers
Simon Becher et al.
Analytical chemistry, 90(19), 11486-11494 (2018-09-11)
Phosphatidylcholines are the major phospholipid component of most eukaryotic cell membranes. Phosphatidylcholines have been shown to actively participate in regulatory and metabolic processes. Dysfunctional metabolic processes have been linked to human disease and can result in altered phosphatidylcholine structural features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service