Skip to Content
Merck
All Photos(2)

Key Documents

850446C

Avanti

14:0-18:0 PC

1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-tetradecanoyl-2-octadecanoyl-sn-glycero-3-phosphocholine; MSPC; PC(14:0/18:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$189.00

About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850446C-25mg)
pkg of 2 × 4 mL (850446C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850446C

concentration

10 mg/mL (850446C-25mg)
25 mg/mL (850446C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-19-20-21-23-25-27-29-31-33-40(43)49-38(37-48-50(44,45)47-35-34-41(3,4)5)36-46-39(42)32-30-28-26-24-22-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3/t38-/m1/s1

InChI key

TYAQXZHDAGZOEO-KXQOOQHDSA-N

General description

Phosphatidylcholine (PC) is one of the most important constituent of colon′s mucosal layer.[1] 14:0-18:0 PC (1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine) is an asymmetrical lipid.[2]

Application

14:0-18:0 PC (1-myristoyl-2-stearoyl-sn-glycero-3-phosphocholine) may be used:
  • in lipid bilayers to study its miscibility with d-erythro-N-palmitoyl-sphingomyelin (16:0-SM) using differential scanning calorimetry (DSC)[3]
  • as an asymmetrical lipid in liposomes to study the impact of bilayer area and lipid geometry on the design of liposomes as liposomal solubilizing agents[2]
  • to prepare a low temperature-sensitive liposome encapsulating thrombolytics to assess thrombolytic activity following hyperthermia[4]

Biochem/physiol Actions

Phosphatidylcholine (PC) can form a hydrophobic surface in the mucus by acting as a surfactant to inhibit the penetrance of bacteria.[1]

Packaging

5 mL Clear Glass Sealed Ampule (850446C-200mg)
5 mL Clear Glass Sealed Ampule (850446C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Vishal Saxena et al.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group, 31(1), 67-73 (2015-03-15)
Clinical efficacy of thrombolytic drugs is limited by lack of specific delivery and requires large therapeutic doses which increase toxicity. Encapsulating these drugs in temperature-sensitive liposomes and applying hyperthermia to deliver thrombolytic agents locally to thrombus might theoretically favourably alter
Bohdana Térová et al.
Biochimica et biophysica acta, 1667(2), 182-189 (2004-12-08)
In this study we have used differential scanning calorimetry (DSC) to study the miscibility of different saturated phosphatidylcholines (PCs) with D-erythro-N-palmitoyl-sphingomyelin (16:0-SM). Information about the miscibility was obtained by observing the thermotropic phase behavior of binary mixtures of saturated PCs
Van Du Nguyen et al.
ACS applied materials & interfaces, 12(9), 10130-10141 (2020-02-12)
Although great efforts have been undertaken to develop a nanoparticle-based drug delivery system (DDS) for the treatment of solid tumors, the therapeutic outcomes are still limited. Immune cells, which possess an intrinsic ability to phagocytose nanoparticles and are recruited by
M Habib Ali et al.
International journal of pharmaceutics, 453(1), 225-232 (2012-07-07)
Liposomes are well recognised for their ability to improve the delivery of a range of drugs. More commonly they are applied for the delivery of water-soluble drugs, but given their structural attributes, they can also be employed as solubilising agents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service