850399P
Avanti
24:1 (Cis) PC
1,2-dinervonoyl-sn-glycero-3-phosphocholine, powder
Synonym(s):
1,2-di-(15Z-tetracosenoyl)-sn-glycero-3-phosphocholine; PC(24:1(15Z)/24:1(15Z)); 110930
About This Item
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 25 mg (850399P-25mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 850399P
lipid type
cardiolipins
phospholipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCC/C=C\CCCCCCCC)=O)=O
InChI
1S/C56H108NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-46-48-55(58)62-52-54(53-64-66(60,61)63-51-50-57(3,4)5)65-56(59)49-47-45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,54H,6-19,24-53H2,1-5H3/b22-20-,23-21-/t54-/m1/s1
InChI key
REWNAIYQZZOESC-CGKZWXATSA-N
General description
Application
Biochem/physiol Actions
Packaging
Legal Information
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service