Skip to Content
Merck
All Photos(2)

Key Documents

850371C

Avanti

22:0 PC

1,2-dibehenoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-didocosanoyl-sn-glycero-3-phosphocholine; PC(22:0/22:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$161.00
500 MG
$463.00

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$161.00
500 MG
$463.00

About This Item

Empirical Formula (Hill Notation):
C52H104NO8P
CAS Number:
Molecular Weight:
902.36
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850371C-25mg)
pkg of 1 × 20 mL (850371C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850371C

concentration

10 mg/mL (850371C-25mg)
25 mg/mL (850371C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OCC[N+](C)(C)C)OCC(OC(=O)CCCCCCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCCCCCC

InChI

1S/C52H104NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-51(54)58-48-50(49-60-62(56,57)59-47-46-53(3,4)5)61-52(55)45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h50H,6-49H2,1-5H3

InChI key

HRTBOPUWPUXROO-UHFFFAOYSA-N

General description

Phosphatidylcholines are phospholipids, that are abundantly present in the membrane. They have fatty acids in the sn-1 and sn-2 positions, that are covalently attached to a glycerol with the help of ester bonds.[1]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

22:0 PC may be used:
  • to study its effects on non-amyloidogenic processing of amyloid precursor protein (APP)/α-secretase activity[2]
  • to prepare a lipid monolayer to study whether ligand is specifically targeted to domains containing Cu(II)-iminodiacetate lipid (Cu-PSIDA)[3]
  • as a lipid standard for the quantification of lipid species from lipid droplets by hydrophilic interaction liquid chromatography-high performance liquid chromatography (HILIC-HPLC) [4]

Biochem/physiol Actions

The physical properties of membranes can be modulated with the help of fatty acyl configuration of phosphatidylcholines. They act as a vital source for eicosanoids.[1]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850371C-500mg)
5 mL Clear Glass Sealed Ampule (850371C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Maloney et al.
Chemistry & biology, 3(3), 185-192 (1996-03-01)
Chemical information is sometimes transmitted across cell membranes by ligand-induced assembly of receptors. We have previously designed a series of lipids with metal-chelating headgroups that can serve as general receptors for proteins containing accessible histidines. Such lipids can also be
Elinor Thompson et al.
Healthcare technology letters, 6(5), 138-142 (2019-12-14)
Oxygen loaded microbubbles are being investigated as a means of reducing tumour hypoxia in order to improve response to cancer therapy. To optimise this approach, it is desirable to be able to measure changes in tissue oxygenation in real-time during
Estelle Beguin et al.
Journal of controlled release : official journal of the Controlled Release Society, 317, 23-33 (2019-11-17)
Recent pre-clinical studies have demonstrated the potential of combining chemotherapy and sonodynamic therapy for the treatment of pancreatic cancer. Oxygen-loaded magnetic microbubbles have been explored as a targeted delivery vehicle for this application. Despite preliminary positive results, a previous study
Pinunta Nittayacharn et al.
Frontiers in pharmacology, 11, 644-644 (2020-06-02)
Drug delivery to solid tumors using echogenic nanobubbles (NBs) and ultrasound (US) has recently gained significant interest. The approach combines attributes of nanomedicine and the enhanced permeation and retention (EPR) effect with the documented benefits of ultrasound to improve tumor
Marcus O W Grimm et al.
International journal of molecular sciences, 14(3), 5879-5898 (2013-03-15)
Alzheimer's disease (AD) is characterized by extracellular accumulation of amyloid-β peptide (Aβ), generated by proteolytic processing of the amyloid precursor protein (APP) by β- and γ-secretase. Aβ generation is inhibited when the initial ectodomain shedding is caused by α-secretase, cleaving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service