Skip to Content
Merck
All Photos(2)

Key Documents

850346C

Avanti

14:1 (Δ9-Cis) PC

1,2-dimyristoleoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(9Z-tetradecenoyl)-sn-glycero-3-phosphocholine; PC(14:1(9Z)/14:1(9Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$346.00

$346.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$346.00

About This Item

Empirical Formula (Hill Notation):
C36H68NO8P
CAS Number:
Molecular Weight:
673.90
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$346.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850346C-25mg)
pkg of 2 × 4 mL (850346C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850346C

concentration

10 mg/mL (850346C-25mg)
25 mg/mL (850346C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) belongs to the class of glycerophospholipid. It is the prime cellular phospholipid.[1]
Phosphatidylcholine (PC) is a strong bilayer-forming lipid. It the most common phospholipid in mammalian membranes.[2] It is also an important component of the mucosal layer of the colon.[3]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

14:1 (Δ9-Cis) PC has been used in the liposome preparation.[4] It has also been used as an internal standard for the extraction of lipids from the interstitial fractions of lung tissues for electrospray ionisation-mass spectrometry (ESI-MS) analysis.[5]

Biochem/physiol Actions

Phosphatidylcholine (PC) contributes to the membrane structure. Each cell type and tissue maintain a unique PC profile. PC plays a key role in cell signaling. Disruption in the homeostasis of PC leads to cell death in mammalian cells.[1] PC is present in bile and aids in lipid emulsification in the diet. It is known to reduce cholesterol and triglyceride levels. It is administered for treating fat embolism and cardiac ischemia.[6] Monounsaturated PC 14:1 (Δ9-Cis) is known to inhibit cell proliferation in different cell lines.[7]
Phosphatidylcholine (PC) functions as a surfactant within the mucus to form a hydrophobic surface to inhibit bacterial penetrance.[3] It is used to treat fat embolism. Phosphatidylcholine lowers the levels of cholesterol and triglycerides.[6]

Packaging

5 mL Clear Glass Sealed Ampule (850346C-200mg)
5 mL Clear Glass Sealed Ampule (850346C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Phosphatidylcholine and cell death
Cui Z and Houweling M
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1585(2-3), 87-96 (2002)
Jiao Cao et al.
Nature communications, 10(1), 5668-5668 (2019-12-13)
Biological nanopores are capable of resolving small analytes down to a monoatomic ion. In this research, tetrachloroaurate(III), a polyatomic ion, is discovered to bind to the methionine residue (M113) of a wild-type α-hemolysin by reversible Au(III)-thioether coordination. However, the cylindrical
Interference of phosphatidylcholines with in vitro cell proliferation?no flock without black sheep
Werlein A, et al.
Biochimica et Biophysica Acta - Biomembranes, 1848(7), 1599-1608 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service