Skip to Content
Merck
All Photos(2)

Key Documents

850310C

Avanti

DOBAQ

Avanti Research - A Croda Brand 850310C

Synonym(s):

N-(4-carboxybenzyl)-N,N-dimethyl-2,3-bis(oleoyloxy)propan-1-aminium

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$284.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$284.00

About This Item

Empirical Formula (Hill Notation):
C49H83NO6
CAS Number:
Molecular Weight:
782.19
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850310C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850310C

concentration

10 mg/mL (850310C-25mg)

lipid type

cationic lipids
headgroup modified lipids

shipped in

dry ice

storage temp.

−20°C

General description

DOBAQ is a cationic lipids with a quaternary amine and unsaturated hydrocarbon chains.[1] It is a pH-sensitive lipid.[2]

Biochem/physiol Actions

N-(4-carboxybenzyl)-N,N-dimethyl2,3-bis(oleoyloxy)propan-1-aminium/DOBAQ displays pH-dependent ionization.[2]

Packaging

5 mL Clear Glass Sealed Ampule (850310C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liposomes in Analytical Methodologies (2016)
Lipid nanoparticles for delivery of messenger RNA to the back of the eye
Patel S, et al.
J. Controlled Release, 303, 91-100 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service