Skip to Content
Merck
All Photos(2)

Key Documents

850158P

Avanti

18:0-20:4 PI(4)P

Avanti Research - A Croda Brand 850158P, powder

Synonym(s):

1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′-phosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$2,030.00
1 MG
$2,800.00

$2,030.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
500 μG
$2,030.00
1 MG
$2,800.00

About This Item

Empirical Formula (Hill Notation):
C47H90 N2O16 P2
CAS Number:
Molecular Weight:
1001.17
UNSPSC Code:
51191904
NACRES:
NA.25

$2,030.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (with stopper and crimp cap (850158P-1mg))
pkg of 1 × 500 μg (with stopper and crimp cap (850158P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850158P

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](OP([O-])(O)=O)[C@@H](O)[C@H]1O)=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C47H84O16P2.2H3N/c1-3-5-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-36-41(49)61-39(37-59-40(48)35-33-31-29-27-25-23-21-18-16-14-12-10-8-6-4-2)38-60-65(57,58)63-47-44(52)42(50)46(43(51)45(47)53)62-64(54,55)56;;/h11,13,17,19,22,24,28,30,39,42-47,50-53H,3-

InChI key

HJHASIMOAXACIC-BZFUBISXSA-N

General description

18:0-20:4 PI(4)P or 1-stearoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′-phosphate) (ammonium salt) is an ammonium salt of phosphoinositol-4-phosphate, with substitution of stearic acid and arachidonic acid at the sn-1 and sn-2 positions of the glycerol backbone, respectively.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850158P-1mg)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850158P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service