Skip to Content
Merck
All Photos(2)

Key Documents

840861C

Avanti

18:0-18:1 PA

1-stearoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1-octadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphate (sodium salt); SOPA; PA(18:0/18:1(9Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$224.00

$224.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$224.00

About This Item

Empirical Formula (Hill Notation):
C39H74O8PNa
CAS Number:
Molecular Weight:
724.96
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$224.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840861C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840861C

concentration

10 mg/mL (840861C-25mg)

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC)O

InChI

1S/C39H75O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,37H,3-17,19,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b20-18-;/t37-;/m1./s1

InChI key

QTWIAVIWKTYOHY-HTNAYUJGSA-M

General description

18:0-18:1 PA (1-stearoyl-2-oleoyl-sn-glycero-3-phosphate) is an unsaturated phosphatidic acid (PA) derivative.[1] PA is present in the biomembranes.[2] It is the simplest glycerophospholipid.[3]

Application

18:0-18:1 PA (1-stearoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt)) may be used:
  • as a phosphatidic acid (PA) standard to identify the corresponding radiolabeled PA on the thin-layer chromatography (TLC)[4]
  • in vesicles for experimental validation in order to identify novel phospholipids capable of inhibiting cellular membrane transportation by (ATP)-binding cassette P-glycoprotein (P-gp)[1]
  • to treat young adult mouse colonic (YAMC) cells prior to nanoclustering- forster resonance energy transfer (FRET) analysis(6)

Biochem/physiol Actions

Phosphatidic acid (PA) is the precursor for other phospholipids and acts as a signaling molecule.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840861C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jolanta Zegarlińska et al.
Acta biochimica Polonica, 65(2), 163-171 (2018-06-19)
Phosphatidic acid (PA) is the simplest glycerophospholipid naturally occurring in living organisms, and even though its content among other cellular lipids is minor, it is drawing more and more attention due to its multiple biological functions. PA is a precursor
Natividad R Fuentes et al.
Cancer research, 78(14), 3899-3912 (2018-05-18)
Ras signaling originates from transient nanoscale compartmentalized regions of the plasma membrane composed of specific proteins and lipids. The highly specific lipid composition of these nanodomains, termed nanoclusters, facilitates effector recruitment and therefore influences signal transduction. This suggests that Ras
Xavier Lucas et al.
PloS one, 8(4), e60679-e60679 (2013-04-18)
P-glycoprotein is capable of effluxing a broad range of cytosolic and membrane penetrating xenobiotic substrates, thus leading to multi-drug resistance and posing a threat for the therapeutic treatment of several diseases, including cancer and central nervous disorders. Herein, a virtual
Y Liu et al.
American journal of physiology. Lung cellular and molecular physiology, 295(3), L471-L478 (2008-07-16)
We have previously reported the participation of mitogen-activated protein, Rho, and phosphoinositide-3 (PI3) kinases in separate pathways in serotonin (5-HT)-induced proliferation of pulmonary artery smooth muscle cells (SMCs). In this study, we investigated the possible participation of phospholipase D (PLD)
Tingting Wang et al.
Frontiers in endocrinology, 9, 221-221 (2018-05-22)
Searching for nutraceuticals and understanding the underlying mechanism that promote fish growth is at high demand for aquaculture industry. In this study, the modulatory effects of soy phosphatidic acids (PA) on cell proliferation, nutrient sensing, and metabolic pathways were systematically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service