840859C
Avanti
16:0-20:4 PA
1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphate (sodium salt), chloroform
Synonym(s):
1-hexadecanoyl-2--(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phosphate (sodium salt); PAPA; PA(16:0/20:4(5Z,8Z,11Z,14Z))
Select a Size
Select a Size
About This Item
Recommended Products
Assay
>99% (TLC)
form
liquid
packaging
pkg of 1 × 2.5 mL (840859C-25mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 840859C
concentration
10 mg/mL (840859C-25mg)
lipid type
phospholipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCC\C=C/C\C=C/C\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)O
InChI
1S/C39H69O8P.Na/c1-3-5-7-9-11-13-15-17-18-19-20-22-24-26-28-30-32-34-39(41)47-37(36-46-48(42,43)44)35-45-38(40)33-31-29-27-25-23-21-16-14-12-10-8-6-4-2;/h11,13,17-18,20,22,26,28,37H,3-10,12,14-16,19,21,23-25,27,29-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b13-11-,18-17-,22-20-,28-26-;/t37-;/m1./s1
InChI key
CHIBHAOESMLHPV-YQVAFVGVSA-M
1 of 4
This Item | 840860C | 840864C | 840887C |
---|---|---|---|
storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
packaging pkg of 1 × 2.5 mL (840859C-25mg) | packaging pkg of 1 × 2.5 mL (840860C-25mg) | packaging pkg of 1 × 2.5 mL (840864C-25mg) | packaging pkg of 1 × 2.5 mL (840887C-25mg) |
form liquid | form liquid | form liquid | form liquid |
shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
manufacturer/tradename Avanti Research™ - A Croda Brand 840859C | manufacturer/tradename Avanti Research™ - A Croda Brand 840860C | manufacturer/tradename Avanti Research™ - A Croda Brand 840864C | manufacturer/tradename Avanti Research™ - A Croda Brand 840887C |
General description
Packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
Target Organs
Central nervous system
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service