Skip to Content
Merck
All Photos(2)

Key Documents

840525P

Avanti

SQDG

Avanti Research - A Croda Brand

Synonym(s):

sulfoquinovosyldiacylglycerol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$629.00
10 MG
$1,010.00

$629.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$629.00
10 MG
$1,010.00

About This Item

Empirical Formula (Hill Notation):
C43H79NO12S
CAS Number:
Molecular Weight:
834.15
UNSPSC Code:
12352211
NACRES:
NA.25

$629.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (840525P-5mg)
pkg of 2 × 5 mg (840525P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCC/C=C\C/C=C\C/C=C\CC)=O)O[C@H](CS(=O)([O-])=O)[C@@H](O)[C@@H]1O.[NH4+]

InChI

1S/C43H76O12S.H3N/c1-3-5-7-9-11-13-15-17-18-20-21-23-25-27-29-31-38(44)52-33-36(34-53-43-42(48)41(47)40(46)37(55-43)35-56(49,50)51)54-39(45)32-30-28-26-24-22-19-16-14-12-10-8-6-4-2;/h5,7,11,13,17-18,36-37,40-43,46-48H,3-4,6,8-10,12,14-16,19-35H2,1-2H3,(H,

InChI key

LCOQSTSHEBORLB-VKQKBWHKSA-N

General description

Sulfoquinovosyl-diacylglycerol (SQDG) is an anionic lipid, which constitutes 20-40% of total lipids in the thylakoid membranes. It is a glycolipid,[1] that plays a major role in photosystem II (PSII) activities in Synechocystis sp. PCC 6803.[2]

Application

SQDG has been used as an additive for photosystems I (PSI) crystallization.[3][4] It has also been used as a standard for mass spectrometry analysis of lipids.[5][6]

Packaging

5 mL Amber Glass Screw Cap Vial (840525P-10mg)
5 mL Amber Glass Screw Cap Vial (840525P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The inhibitory action of SQDG (sulfoquinovosyl diacylglycerol) from spinach on Cdt1-geminin interaction
Mizushina Y, et al.
Biochimie, 90(6), 947-956 (2008)
Crystal structure of photosystem I monomer from Synechocystis PCC 6803
Netzer-El SY, et al.
Frontiers in Plant Science, 9(9), 645-654 (2018)
Sulfoquinovosyldiacylglycerol has an essential role in Thermosynechococcus elongatus BP-1 under phosphate-deficient conditions
Endo K, et al.
Plant & Cell Physiol., pcw159-pcw159 (2016)
Structure and function of wild-type and subunit-depleted photosystem I in Synechocystis
Malavath T, et al.
Biochimica et Biophysica Acta - Bioenergetics, 1859(9), 645-654 (2018)
Transcriptomic and microRNAomic profiling reveals multi-faceted mechanisms to cope with phosphate stress in a dinoflagellate
Shi X, et al.
The ISME Journal, 11(10), 2209-2209 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service