Skip to Content
Merck
All Photos(3)

Key Documents

840074P

Avanti

Soy PA

Avanti Research - A Croda Brand 840074P, powder

Synonym(s):

L-α-phosphatidic acid (Soy) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$761.00

$761.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$761.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$761.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840074P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840074P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C37H69O8P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(39)45-35(34-44-46(40,41)42)33-43-36(38)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,35H,3-10,12,14-16,19-34H2,1-2H3,(H2,40,41,42);/q;+1/p-1/b13-11-,18-17-;/t35-;/m1./s1

InChI key

IYNPOFCKGPJAFU-HQFKUYMNSA-M

General description

Phosphatidic acid (PA) is a diacyl-glycerophospholipid, with two fatty acids and a phosphate group that are covalently attached to a glycerol molecule through ester linkages.[1] PA is synthesized by the activity of the endogenous enzyme phospholipase D or by supplementation with soy lecithin.[2]

Application

Soy PA is suitable for use to study its effect on cell proliferation, nutrient sensing and to examine metabolic pathways in primary muscle cells of turbot (Scophthalmus maximus).[2]

Biochem/physiol Actions

Phosphatidic acid (PA) is a prime glycerophospholipid present in the biomembrane and is a precursor for phosphatidylcholine or phosphatidylserine. It has different functions in the cells like influencing the membrane curvature and signaling.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840074P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

The mitotic and metabolic effects of phosphatidic acid in the primary muscle cells of turbot (Scophthalmus maximus)
Wang T, et al.
Frontiers in Endocrinology, 9 (2018)
Phosphatidic acid enhances mTOR signaling and resistance exercise induced hypertrophy
Joy JM, et al.
Nutrition, 11(1), 29-29 (2014)
Daniel Schlam et al.
The Journal of biological chemistry, 288(32), 23090-23104 (2013-07-03)
It is commonly assumed that all phagosomes have identical molecular composition. This assumption has remained largely unchallenged due to a paucity of methods to distinguish individual phagosomes. We devised an assay that extends the utility of nitro blue tetrazolium for
Michal Bohdanowicz et al.
Molecular biology of the cell, 24(11), 1700-1712 (2013-04-12)
Macrophages and dendritic cells continuously survey their environment in search of foreign particles and soluble antigens. Such surveillance involves the ongoing extension of actin-rich protrusions and the consequent formation of phagosomes and macropinosomes. The signals inducing this constitutive cytoskeletal remodeling
Chi-Lun Chang et al.
The Journal of biological chemistry, 290(23), 14289-14301 (2015-04-19)
Phosphatidylinositol (PI) 4,5-bisphosphate (PIP2) at the plasma membrane (PM) constitutively controls many cellular functions, and its hydrolysis via receptor stimulation governs cell signaling. The PI transfer protein Nir2 is essential for replenishing PM PIP2 following receptor-induced hydrolysis, but key mechanistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service