Skip to Content
Merck
All Photos(2)

Key Documents

840066C

Avanti

20:4 PS

1,2-diarachidonoyl-sn-glycero-3-phospho-L-serine (sodium salt), chloroform

Synonym(s):

1,2-di-(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); DAPS; PS(20:4(5Z,8Z,11Z,14Z)/20:4(5Z,8Z,11Z,14Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H73NO10PNa
CAS Number:
Molecular Weight:
854.04
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840066C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840066C

concentration

10 mg/mL (840066C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O.[Na+]

InChI

1S/C46H74NO10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-44(48)54-39-42(40-55-58(52,53)56-41-43(47)46(50)51)57-45(49)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h11-14,17-20,23-26,29-32,42-43H,3-10,15-16,21-22,27-28,33-41,47H2,1-2H3,(H,50,51)(H,52,53);/q;+1/p-1/b13-11-,14-12-,19-17-,20-18-,25-23-,26-24-,31-29-,32-30-;/t42-,43+;/m1./s1

InChI key

JFPYEVXNLNLLRD-LAECUUORSA-M

General description

20:4 PS or 1,2-diarachidonoyl-sn-glycero-3-phospho-L-serine is a sodium salt of glycerophospholipid having two chains of arachidonic acid attached to the sn-1 and sn-2 positions and phospho-L-serine at the sn-3 position of the glycerol backbone.

Application

20:4 PS or 1,2-diarachidonoyl-sn-glycero-3-phospho-L-serine (sodium salt) has been used in liposome preparation to study effects of the fatty acid (FA) profile of phospholipids on membrane vesiculation by dynamin and endophilin.[1]

Packaging

5 mL Clear Glass Sealed Ampule (840066C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco M Manni et al.
eLife, 7 (2018-03-16)
Phospholipid membranes form cellular barriers but need to be flexible enough to divide by fission. Phospholipids generally contain a saturated fatty acid (FA) at position sn1 whereas the sn2-FA is saturated, monounsaturated or polyunsaturated. Our understanding of the impact of
Peter A Leventis et al.
Annual review of biophysics, 39, 407-427 (2010-03-03)
Phosphatidylserine (PS) is the most abundant negatively charged phospholipid in eukaryotic membranes. PS directs the binding of proteins that bear C2 or gamma-carboxyglutamic domains and contributes to the electrostatic association of polycationic ligands with cellular membranes. Rather than being evenly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service