Skip to Content
Merck
All Photos(1)

Key Documents

810508P

Avanti

16:0 DNP PE

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(2,4-dinitrophenyl) (ammonium salt), powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-(2,4-dinitrophenyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$284.00

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$284.00

About This Item

Empirical Formula (Hill Notation):
C43H79N4O12P
CAS Number:
Molecular Weight:
875.08
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$284.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (810508P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810508P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphoethanolamine is a precursor to phosphatidylcholine and phosphatidylethanolamine.[1] 2,4-dinitrophenol (DNP) is a synthetic lipid-soluble compound which transports ions across the bioenergetic membranes.[2]

Application

16:0 DNP PE may be used:
  • to generate lipid aggregates/lipid micelles[3]
  • in the preparation of liposomes[4]
  • as an internal standard to spike the tissue samples for the quantification of N-acylphosphatidylethanolamine (NAPE)[5]
  • in lipid solution as a receptor site to prepare bilayer lipid membranes (BLMs)[6]

Biochem/physiol Actions

Phosphoethanolamine participates in phospholipid metabolism.[7] It has the ability to repress tumour growth both in vitro and in vivo.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810508P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesse LoVerme et al.
The Journal of biological chemistry, 281(32), 22815-22818 (2006-06-21)
Oleoylethanolamide (OEA) is an endogenous lipid mediator that inhibits feeding and stimulates lipolysis by activating the nuclear receptor peroxisome proliferator-activating receptor-alpha. Little is known about the physiological regulation of this compound outside of the gastrointestinal tract, where its production is
Membrane Transport
An Introduction to Biological Membranes, 423-451 (2016)
Anna Ketelsen et al.
Journal of immunological methods, 339(2), 195-204 (2008-10-16)
Antibodies against hydrophobic antigens are common in several autoimmune diseases. However, detection of such antibodies by standard immune-assays, such as ELISA, is problematic, in part because of the problems with coating hydrophobic molecules onto polystyrene multi-well plates. We describe a
Mieko Horie et al.
Analytical biochemistry, 369(2), 192-201 (2007-08-28)
This article describes a new method for direct fluorometric immunoassay with a liposome array using pH-sensitive dye (BCECF [2',7'-bis(carboxyethyl)-4 or 5-carboxyfluorescein])-encapsulating liposomes immobilized on an avidin slip and gramicidin channels. The liposomes were composed of phosphatidylcholine (PC), cholesterol (Chol), biotinylated
Ayumi Hirano et al.
Biosensors & bioelectronics, 18(8), 973-983 (2003-06-05)
A novel ion-channel sensor based on a membrane bound receptor and a single gramicidin channel is described, in which the binding of an analyte to the membrane bound receptor modulates the single-channel activity of gramicidin. The sensor is composed of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service