Skip to Content
Merck
All Photos(1)

Key Documents

810382P

Avanti

18:1 TopFluor PE

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[(dipyrromethaneboron difluoride)butanoyl], powder

Synonym(s):

18:1 TopFluor PE

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$569.00
10 MG
$3,090.00

$569.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$569.00
10 MG
$3,090.00

About This Item

Empirical Formula (Hill Notation):
C58H100BF2N4O9P
CAS Number:
Molecular Weight:
1077.22
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$569.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810382P-1mg)
pkg of 1 × 10 mg (810382P-10mg)
pkg of 1 × 5 mg (810382P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand (810382P)

shipped in

dry ice

storage temp.

−20°C

General description

TopFluor is dipyrrometheneboron difluoride undecanoic acid, used for labelling lipids. Fluorescent lipids find its application in live imaging in animal model and in cell culture. TopFluor is a green variant of boron-dipyrromethene (BODIPY) (a fluorescent tag for lipids). TopFluor compared to BODIPY, possesses two additional methyl groups and the lipid conjugate is bonded to carbon 1 of its central ring. It forms a T-shaped molecule in contrast to the linear molecule formed by BODIPY FL- fatty acids FAs).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810382P-10mg)
5 mL Amber Glass Screw Cap Vial (810382P-1mg)
5 mL Amber Glass Screw Cap Vial (810382P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An HPLC-CAD/fluorescence lipidomics platform using fluorescent fatty acids as metabolic tracers
Quinlivan VH, et al.
Journal of Lipid Research, 58(5), 1008-1020 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service