Skip to Content
Merck
All Photos(1)

Key Documents

800854P

Avanti

1,3-17:0 D5 DG

Avanti Research - A Croda Brand, powder

Synonym(s):

1,3-Diheptadecanoyl-glycerol (d5)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$511.00

$511.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$511.00

About This Item

Empirical Formula (Hill Notation):
C37H67D5O5
CAS Number:
Molecular Weight:
602.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$511.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (800854P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OC(CCCCCCCCCCCCCCCC)=O)(O)C([2H])([2H])OC(CCCCCCCCCCCCCCCC)=O

InChI

1S/C37H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36(39)41-33-35(38)34-42-37(40)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h35,38H,3-34H2,1-2H3/i33D2,34D2,35D

InChI key

WDSOLIGXMKFAGU-WJZUKHFDSA-N

Application

1,3-17:0 D5 DG has been used as an internal standard for lipid extraction.[1] It has also been used for the incubation of rat tissue samples.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (800854P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional Effects of the Buckwheat Iminosugar d-Fagomine on Rats with Diet-Induced Prediabetes
Ramos-Romero S, et al.
Molecular Nutrition And Food Research, 62(16), 1800373-1800373 (2018)
Sara Ramos-Romero et al.
Molecular nutrition & food research, 64(1), e1900564-e1900564 (2019-10-28)
This study examines the long-term functional effects of d-fagomine on sucrose-induced factors of metabolic dysfunctions and explores possible molecular mechanisms behind its action. Wistar Kyoto rats are fed a 35% sucrose solution with d-fagomine (or not, for comparison) or mineral
MIGA2 Links Mitochondria, the ER, and Lipid Droplets and Promotes De Novo Lipogenesis in Adipocytes
Freyre C A C, et al.
Molecular Cell, 76(5), 811-825 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service