Skip to Content
Merck
All Photos(1)

Key Documents

791297P

Avanti

bis(14:0 PE)-DTPA (Gd)

Avanti Research - A Croda Brand 791297P, powder

Synonym(s):

bis(1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine)-N-N′-diethylenetriaminepentaacetic acid (gadolinium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$225.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$225.00

About This Item

Empirical Formula (Hill Notation):
C80H154GdN7O24P2
CAS Number:
Molecular Weight:
1817.31
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (791297P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 791297P

shipped in

dry ice

storage temp.

−20°C

General description

bis(14:0 PE)-DTPA (Gd) is a lipidic gadolinium complex,[1] with four units of myristic acid. The diethylenetriaminepentaacetic acid is chelated to gadolinium ion.

Application

bis(14:0 PE)-DTPA (Gd) has been used in liposome preparation.[1]
bis(14:0 PE)-DTPA (Gd)(bis(1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine)-N-N′-diethylenetriaminepentaacetic acid (gadolinium salt)) has been used in liposome preparation for magnetic resonance imaging (MRI) measurements.[1]

Biochem/physiol Actions

Gadolinium based lipid complexes are contrast agents in magnetic resonance imaging. Such lipid complexes are used in liposome-based bilayer preparation for imaging.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (791297P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multifunctional receptor-targeted nanocomplexes for magnetic resonance imaging and transfection of tumours
Kenny GD, et al.
Biomaterials, 33(29), 7241-7250 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service