Skip to Content
Merck
All Photos(1)

Key Documents

790304P

Avanti

Coag Reagent I

Avanti Research - A Croda Brand 790304P, powder

Synonym(s):

Coag Reagent, Coagulation Reagent I, contains DOPE:DOPS:DOPC (5:3:2 w/w)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$487.00
200 MG
$853.00

$487.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$487.00
200 MG
$853.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

$487.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 2 × 100 mg (790304P-200mg)
pkg of 1 × 25 mg (790304P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790304P

application(s)

microbiology

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Coag Reagent I comprises 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine(DOPE), 1,2-dioleoyl-sn-glycero-3-phospho-L-serine (DOPS) and 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) in the ratio 5:3:2 w/w. They are present predominantly in the synaptic vesicle membrane.[1] DOPC has choline group and DOPE comprises ethanolamine moiety. At neutral pH, DOPE and DOPC exist as a zwitterion whereas DOPS takes up a negative charge and has a serine group.[2]

Application

Coag Reagent I has been used in the preparation of small unilamellar vesicles (SUVs) for α-synuclein binding studies.[3][1] It may be used in the preparation of bilayers for reconstituting ryanodine receptor type 2 (RyR2) for single-channel recording studies.[4] It may be used for glass supported lipid bilayers preparation.[2]

Biochem/physiol Actions

Coag Reagent I mixed lipid is useful in the preparation of supported lipid bilayers and platelet membranes for coagulation studies.[2] It is used in the small unilamellar vesicles (SUVs) generation for α-synuclein and squalamine binding studies.[1]

Packaging

5 mL Clear Glass Sealed Ampule (790304P-200mg)
5 mL Clear Glass Sealed Ampule (790304P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

EL20, a potent antiarrhythmic compound, selectively inhibits calmodulin-deficient ryanodine receptor type 2
Klipp RC, et al.
Heart Rhythm : the Official Journal of the Heart Rhythm Society, 15(4), 578-586 (2018)
A natural product inhibits the initiation of alpha-synuclein aggregation and suppresses its toxicity
Perni M, et al.
Proceedings of the National Academy of Sciences of the USA, 114(6), E1009-E1017 (2017)
Three-Dimensional Heterogeneous Structure Formation on a Supported Lipid Bilayer Disclosed by Single-Particle Tracking
Zhong Y & Wang G
Langmuir, 34(39), 11857-11865 (2018)
A M van den Besselaar et al.
Clinical chemistry, 43(7), 1215-1222 (1997-07-01)
To contribute to the development of a reference reagent for monitoring heparin therapy, a lyophilized partial thromboplastin time (PTT) reagent was prepared from synthetic dioleoylphosphatidylcholine, dioleoylphosphatidylserine, and dioleoylphosphatidylethanolamine, with colloidal silica as activator. The reagent, coded 91/558, was contained in
A M van den Besselaar et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 4(6), 895-903 (1993-12-01)
The activated partial thromboplastin time (aPTT) is the most popular test for monitoring of heparin therapy. The purpose of the present study was to show that an aPTT reagent with good response to heparin can be prepared from synthetic phosphoglycerides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service