Skip to Content
Merck
All Photos(1)

Key Documents

700299O

Avanti

Farnesol

Avanti Research - A Croda Brand

Synonym(s):

(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$192.00

$192.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$192.00

About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$192.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

oil

packaging

package of 1 × 25 mg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C

InChI key

CRDAMVZIKSXKFV-YFVJMOTDSA-N

Packaging

5 mL Clear Glass Sealed Ampule (700299O-25MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service