Skip to Content
Merck
All Photos(1)

Documents

700226L

Avanti

22:4 Cholesteryl-d7 ester

Avanti Research - A Croda Brand

Synonym(s):

cholesteryl-d7 docosa-7,10,13,16-tetraenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H73D7O2
CAS Number:
Molecular Weight:
708.20
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

>99% (TLC)

form

solution

packaging

package of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Packaging

2 mL Amber Glass Sealed Ampule (700226L-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service