Skip to Content
Merck
All Photos(1)

Key Documents

700158P

Avanti

Sitoindoside I

6-O-palmitoyl-sitosteryl-β-D-glucose, powder

Synonym(s):

3-O-[(6′-O-palmitoyl)-β-D-glucopyranosyl] sitosterol; 16:0 sitosteryl glucose

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$224.00

$224.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$224.00

About This Item

Empirical Formula (Hill Notation):
C51H90 O7
CAS Number:
Molecular Weight:
815.26
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$224.00


Estimated to ship onMarch 30, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700158P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700158P

shipped in

dry ice

storage temp.

−20°C

General description

Sitoindoside I is an acyl steryl glycoside.[1]
Sitoindoside I is present in fruits like banana (Musa paradisiaca).[2] It is an acyl steroid glycoside phytochemical.[3] Sitoindoside I has heterogeneous aglycon composition.[4] It is hypoglycemic agent.[5]
Sitoindosides are natural sterols, containing ergostane. It holds a glucose moiety at carbon 27.[6]

Biochem/physiol Actions

Plant based sterylglucosides have neurotoxic functionality and are correlated to the pathogenesis of neurodegenerative disorders in vivo.[4]
Sitoindoside I is known to exhibit anti ulcerogenic action.[7]

Packaging

5 mL Amber Glass Screw Cap Vial (700158P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Musa paradisiaca L. and Musa sapientum L.: a phytochemical and pharmacological review
Imam MZ and Akter S
Journal of Applied Pharmaceutical Science, 1(5), 14-20 null
Fruit and vegetables: harvesting, handling and storage (2014)
Fruit and vegetables: harvesting, handling and storage (2008)
Musa paradisiaca L. and Musa sapientum L.: a phytochemical and pharmacological review
Imam MZ and Akter S
Journal of Applied Pharmaceutical Science, 1(5), 14-20 (2011)
Fruit and Vegetables: Harvesting, Handling and Storage (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service