Skip to Content
Merck
All Photos(1)

Key Documents

700138P

Avanti

7-dehydrodesmosterol

powder

Synonym(s):

Cholesta-5,7,24-trien-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,260.00

$1,260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,260.00

About This Item

Empirical Formula (Hill Notation):
C27H42O
CAS Number:
Molecular Weight:
382.62
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (700138P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700138P

shipped in

dry ice

storage temp.

−70°C

SMILES string

O[C@H]1CC[C@@]2([C@H]3CC[C@]4([C@@H](CC[C@@H]4[C@@H](CCC=C(C)C)C)C3=CC=C2C1)C)C

InChI

1S/C27H42O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9-10,19,21,23-25,28H,6,8,11-17H2,1-5H3/t19-,21+,23-,24+,25+,26+,27-/m1/s1

InChI key

RUSSPKPUXDSHNC-DDPQNLDTSA-N

General description

7-dehydrodesmosterol (7-DHD) is synthesized via the Bloch pathway. 7-DHD serves as a desmosterol synthesis precursor and the enzyme 7-dehydrocholesterol reductase (DHCR7) catalyzes this step.[1]

Application

7-dehydrodesmosterol has been used as an internal oxysterol standard in ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS).[2]

Biochem/physiol Actions

7-dehydrodesmosterol (7-DHD) levels are higher during fetal development[3] and increases with aging.[4] Elevated levels of 7-DHD is associated with Smith-Lemli-Opitz syndrome mouse model.[3] Reduced levels of 7-DHD is observed in shiverer and quaking disorder.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (700138P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J M Bourre et al.
Journal of neurochemistry, 54(4), 1196-1199 (1990-04-01)
In rat sciatic nerve, the 7-dehydrocholesterol content decreased dramatically during the postnatal period and slowly during adulthood and aging. In contrast, the 7-dehydrodesmosterol content peaked at 14 days and was nearly undetectable after 60 days. The desmosterol content peaked at
Alterations of cholesterol synthesis precursors (7-dehydrocholesterol, 7-dehydrodesmosterol, desmosterol) in dysmyelinating neurological mutant mouse (quaking, shiverer and trembler) in the PNS and the CNS
Bourre JM, et al.
Biochimica et Biophysica Acta, Lipids and Lipid Metabolism, 1004(3), 387-390 null
Anna Meljon et al.
Biochemical pharmacology, 86(1), 43-55 (2013-03-19)
In this study the sterol and oxysterol profile of newborn brain from the Dhcr7(Δ3-5/T93M) mouse model of Smith-Lemli-Opitz syndrome (SLOS) has been investigated. This is a viable mouse model which is compound heterozygous containing one null allele and one T93M
Josi Herron et al.
Current protocols in toxicology, 78(1), e65-e65 (2018-10-16)
Cholesterol and cholesterol-derived oxysterols are critical for embryonic development, synapse formation and function, and myelination, among other biological functions. Indeed, alterations in levels of cholesterol, sterol precursors, and oxysterols result in a variety of developmental disorders, emphasizing the importance of
Vladimiro Cardenia et al.
Biochimie, 153, 99-108 (2018-08-06)
The electronic cigarettes (e-cigarettes, e-cigs) have become the most sought-after alternative to the traditional cigarettes, partly due to the widespread perception of safety. However, the high temperature reached by e-cig solutions can generate toxic compounds, some of which are listed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service