Skip to Content
Merck
All Photos(2)

Key Documents

700052P

Avanti

22(R)-hydroxycholesterol-d7

Avanti Research - A Croda Brand

Synonym(s):

25,26,26,26,27,27,27-heptadeuterocholest-5-ene-3β,22R-diol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,260.00

$1,260.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$1,260.00

About This Item

Empirical Formula (Hill Notation):
C27H39O2D7
CAS Number:
Molecular Weight:
409.70
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

$1,260.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

description

cholest-5-ene-3β,22(R)-diol-d7

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700052P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

General description

22(R)-hydroxycholesterol is a diastereomer of 22(S)-hydroxycholesterol.[1] It is present in the neonate brain.[2] 22(R)-hydroxycholesterol-d7 is a deuterated form of 22(R)-hydroxycholesterol.

Biochem/physiol Actions

22(R)-hydroxycholesterol (22(R)-HC) is a liver X receptor (LXR) ligand and is a key for the receptor activation.[3]. 22(R)-HC promotes mesenchymal stem cell osteogenesis along with other oxysterols.[4] It also regulates cholesterol homeostasis[5] and suppresses prostate tumor progression.[6] Low levels of 22(R)-HC is observed in Alzheimer′s disease and may be implicated in neuroinflammation and neurodegenerative diseases.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700052P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan M Augustin et al.
The Plant journal : for cell and molecular biology, 82(6), 991-1003 (2015-05-06)
Steroid alkaloids have been shown to elicit a wide range of pharmacological effects that include anticancer and antifungal activities. Understanding the biosynthesis of these molecules is essential to bioengineering for sustainable production. Herein, we investigate the biosynthetic pathway to cyclopamine
Combined treatment with 9-cis beta-carotene and 22R-hydroxycholesterol augments cholesterol efflux in macrophages
Mahler L, et al.
Algal research, 44, 101700-101700 (2019)
Hyunmi Kim et al.
Biochimica et biophysica acta, 1859(8), 1056-1070 (2016-05-22)
MAP kinase phosphatase (MKP)-1 plays a pivotal role in controlling MAP kinase (MAPK)-dependent (patho) physiological processes. Although MKP-1 gene expression is tightly regulated at multiple levels, the underlying mechanistic details remain largely unknown. In this study, we demonstrate that MKP-1
Jin-Sun Lee et al.
Stem cell research & therapy, 8(1), 276-276 (2017-12-07)
Oxysterols, oxygenated by-products of cholesterol biosynthesis, play roles in various physiological and pathological systems. However, the effects of oxysterols on periodontal regeneration are unknown. This study investigated the effects of the specific oxysterol combination of 22(S)-hydroxycholesterol and 20(S)-hydroxycholesterol (SS) on
Chih-Pin Chuu et al.
Anticancer research, 30(9), 3643-3648 (2010-10-15)
Previously, we and other groups reported that liver X receptor (LXR) agonists T0901317, 22(R)-hydroxycholesterol, and 24(S)-hydroxycholesterol suppressed the proliferation of prostate and breast cancer cells. In this study, we report that T0901317 and 22(R)-hydroxycholesterol treatment inhibited the proliferation of different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service