Skip to Content
Merck
All Photos(2)

Key Documents

700039P

Avanti

24(S),25-epoxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

(24S,25)-Epoxycholesterol, (3β,24S)-24,25-Epoxycholest-5-en-3-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,260.00
5 MG
$4,950.00

$1,260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$1,260.00
5 MG
$4,950.00

About This Item

Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 1 mg (700039P-1mg)
pkg of 1 × 5 mg (700039P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O1[C@H](C1(C)C)CC[C@H]([C@@H]2[C@@]3([C@H]([C@H]4[C@@H]([C@]5(CC[C@@H](CC5=CC4)O)C)CC3)CC2)C)C

InChI

1S/C27H44O2/c1-17(6-11-24-25(2,3)29-24)21-9-10-22-20-8-7-18-16-19(28)12-14-26(18,4)23(20)13-15-27(21,22)5/h7,17,19-24,28H,6,8-16H2,1-5H3/t17-,19+,20+,21-,22+,23+,24+,26+,27-/m1/s1

InChI key

OSENKJZWYQXHBN-XVYZBDJZSA-N

General description

24(S),25-epoxycholesterol is an oxysterol, which is produced by fibroblasts, macrophages and brain cells.
24(S),25-epoxycholesterol is a cholesterol-derived oxysterol, which is produced by fibroblasts, macrophages, and the brain cells. 24(S),25-epoxycholesterol is synthesized de novo in the mevalonate pathway.

Application

24(S),25-epoxycholesterol has been used as an oxysterol/internalstandard to study the effects of AY9944-induced changes in neurospheres.

Biochem/physiol Actions

Exogenous addition of 24(S),25-epoxycholesterol lowers 3-hydroxy-3-methylglutaryl-Coenzyme A (HMG-CoA) reductase activity, represses sterol regulatory element-binding protein (SREBP) processing, and induces liver × receptor (LXR) -gene targets in vitro.

Packaging

5 mL Amber Glass Screw Cap Vial (700039P-1mg)
5 mL Amber Glass Screw Cap Vial (700039P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service