Skip to Content
Merck
All Photos(2)

Key Documents

700017P

Avanti

25-hydroxycholesterol-3-sulfate

Avanti Research - A Croda Brand

Synonym(s):

5-cholesten-3beta, 25-diol 3-sulfate; 25HC3S

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$549.00
10 MG
$820.00

$549.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$549.00
10 MG
$820.00

About This Item

Empirical Formula (Hill Notation):
C27H45NaO5S
CAS Number:
Molecular Weight:
504.70
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$549.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

25-hydroxycholesterol-3-sulfate (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (700017P-10mg)
pkg of 1 × 5 mg (700017P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@]12[C@]([C@](CC[C@H](OS(=O)([O-])=O)C3)(C)C3=CC2)([H])CC[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCCC(C)(O)C.[Na+]

InChI key

OEGAOHNRCSZIRO-KSGNISEOSA-M

General description

25-hydroxycholesterol-3-sulfate (25OHC3S) is a nuclear oxysterol.[1] It accumulates in hepatocyte nuclei upon steroidogenic acute regulatory protein (STARD1) overexpression.[2]

Application

25-hydroxycholesterol-3-sulfate has been used to induce anti-inflammatory responses and understand its effect on lymphocyte migration.[3] It has also been used as an oxysterol sulfate standard in mass spectroscopy and liquid chromatography methods.[4]

Biochem/physiol Actions

25-hydroxycholesterol-3-sulfate (25OHC3S) is involved in lipid homeostasis in liver[5] and is a lipid metabolism regulator.[1] 25OHC3S acts on liver X receptor (LXR) and sterol regulatory element-binding protein (SREBP) and inhibits cholesterol synthesis by downregulating enzymes of the biosynthesis pathway in hepatocytes and macrophages.[5] It also acts an agonist for peroxisome proliferator-activated receptor γ (PPAR-γ) and modulates the nuclear factor κ-light-chain-enhancer (NF-κB) based inflammatory response.[2] 25OHC3S modulates tumor necrosis factor α (TNFα)-induced inflammatory response in hepatocytes.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700017P-10mg)
5 mL Amber Glass Screw Cap Vial (700017P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leyuan Xu et al.
American journal of physiology. Endocrinology and metabolism, 302(7), E788-E799 (2012-01-26)
The nuclear receptor peroxisome proliferator-activated receptors (PPARs) are important in regulating lipid metabolism and inflammatory responses in macrophages. Activation of PPARγ represses key inflammatory response gene expressions. Recently, we identified a new cholesterol metabolite, 25-hydroxycholesterol-3-sulfate (25HC3S), as a potent regulatory
Shunlin Ren et al.
Biochemical and biophysical research communications, 360(4), 802-808 (2007-07-13)
Recently, a novel oxysterol, 5-cholesten-3beta, 25-diol 3-sulfate (25HC3S) was identified in primary rat hepatocytes following overexpression of the cholesterol transport protein, StarD1. This oxysterol was also detected in human liver nuclei. In the present study, 25HC3S was chemically synthesized. Addition
Leyuan Xu et al.
Lipids, 45(9), 821-832 (2010-08-12)
Dysregulation of lipid metabolism is frequently associated with inflammatory conditions. The mechanism of this association is still not clearly defined. Recently, we identified a nuclear oxysterol, 25-hydroxycholesterol-3-sulfate (25HC3S), as an important regulatory molecule involved in lipid metabolism in hepatocytes. The
Yongjie Ma et al.
American journal of physiology. Endocrinology and metabolism, 295(6), E1369-E1379 (2008-10-16)
The oxysterol receptor LXR is a key transcriptional regulator of lipid metabolism. LXR increases expression of SREBP-1, which in turn regulates at least 32 genes involved in lipid synthesis and transport. We recently identified 25-hydroxycholesterol-3-sulfate (25HC3S) as an important regulatory
Tetsuya Sakurai et al.
Science signaling, 11(541) (2018-08-02)
Although immune responses are essential to protect the body from infection, they can also harm tissues. Certain tissues and organs, including the eye, constitute specialized microenvironments that locally inhibit immune reactivity. Dedicator of cytokinesis protein 2 (DOCK2) is a Rac-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service