Skip to Content
Merck
All Photos(3)

Key Documents

699852P

Avanti

3D-PHAD

Avanti Research - A Croda Brand

Synonym(s):

Monophosphoryl 3-Deacyl Lipid A (Synthetic) Pat No. 9,241,988

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$338.00
5 MG
$1,350.00

$338.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$338.00
5 MG
$1,350.00

About This Item

Empirical Formula (Hill Notation):
C82H158N3O20P
CAS Number:
Molecular Weight:
1537.11
UNSPSC Code:
12352211
NACRES:
NA.25

$338.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (HPLC)

form

powder

packaging

pkg of 1 × 1 mg (699852P-1mg)
pkg of 1 × 5 mg (699852P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

vaccine development

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]([C@@H](CO[C@@H]([C@@H]1NC(C[C@H](OC(CCCCCCCCCCCCC)=O)CCCCCCCCCCC)=O)O[C@H](CO)[C@@H](OP([O-])(O)=O)[C@@H]1OC(C[C@H](OC(CCCCCCCCCCCCC)=O)CCCCCCCCCCC)=O)O[C@H](O)[C@@H]2NC(C[C@H](O)CCCCCCCCCCC)=O)[C@@H]2O.[NH4+]

InChI

1S/C82H155N2O20P.H3N/c1-6-11-16-21-26-31-33-38-43-48-53-58-72(89)99-66(56-51-46-41-36-29-24-19-14-9-4)61-71(88)84-76-80(103-74(91)62-67(57-52-47-42-37-30-25-20-15-10-5)100-73(90)59-54-49-44-39-34-32-27-22-17-12-7-2)79(104-105(95,96)97)68(63-85)102-82(76)9

InChI key

XERJCJBCCDONJL-WOBTUNBLSA-N

General description

Vaccination is well-accepted as an effective method to prevent infections by mounting pathogen-specific immune responses prior to the infection. Usually, immunization with vaccine antigens alone is not able to induce robust or long-lasting immune responses — resulting in failure of protective immunity against infections. Thus, adjuvants are required to enhance cellular or humoral immune responses upon immunization. Because vaccine adjuvants using Lipid A have proven to be safe and effective in inducing Th-1 type immune responses to heterologous proteins in animal and human vaccines, Avanti developed Phosphorylated HexaAcyl Disaccharide (PHAD), the first fully synthetic monophosphoryl Lipid A available for use as an adjuvant in human vaccines.

Application

3D-PHAD has been used:
  • as a synthetic adjuvant for respiratory syncytial virus (RSV) vaccine[1]
  • as a synthetic adjuvant to prepare hepatitis B virus (HBV) and virus-like particles (VLP)-based human papillomavirus (HPV) vaccines[2]
  • in liposome preparation for the generation of army liposome formulation (ALF) for human immunodeficiency (HIV) gp140 antigen immunization studies in mice.[3]

Biochem/physiol Actions

3-deacyl-phosphorylated hexa-acyl disaccharide (3D-PHAD) is derived from monophosphoryl lipid A. It is amphiphilic and associates with the virosomal membranes.[1]

Packaging

2 mL Amber Glass Crimp Cap Vial (699852P-1mg)
2 mL Amber Glass Crimp Cap Vial (699852P-5mg)

Other Notes

For R&D use only. Not for drug, household, or other uses.

Legal Information

3D-PHAD is a trademark of Avanti Polar Lipids, LLC
Avanti Research is a trademark of Avanti Polar Lipids, LLC
PHAD is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monica L Martin et al.
Malaria journal, 18(1), 377-377 (2019-11-30)
Indian-origin rhesus (InR) are preferred for research, but strict export restrictions continue to limit their use. Chinese-origin rhesus (ChR), although easier to procure, are genetically distinct from InR and differ in their immune response to infectious agents, such as the
Immune response to antigen adsorbed to aluminum hydroxide particles: Effects of co-adsorption of ALF or ALFQ adjuvant to the aluminum-antigen complex
Beck Z, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 275, 1219-1219 (2018)
Immunogenicity of HPV and HBV vaccines: adjuvanticity of synthetic analogs of monophosphoryl lipid A combined with aluminum hydroxide
Taleghani N, et al.
Acta Pathologica, Microbiologica et Immunological Scandinavica, 127(3), 150-157 (2019)
Development of a virosomal RSV vaccine containing 3D-PHAD Adjuvant: formulation, composition, and long-term stability
Lederhofer J
Pharmaceutical Research, 35(9), 172-172 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service