Skip to Content
Merck
All Photos(1)

Key Documents

690050P

Avanti

Soy Phospholipid Mixture

Avanti Research - A Croda Brand

Synonym(s):

Soy Phospholipid Mixture, Phospholipids from Soybean, Contains Soy PC (3.8mg) Soy PE (3.0mg) Soy PI (1.8mg) Soy PA (0.7mg) Soy LPC (0.7mg) with 0.1% BHT

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$324.00

$324.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$324.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

$324.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (HPLC)

form

powder

packaging

pkg of 1 × 10 mg (690050P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

NMR: suitable

lipid type

phospholipids
lipid extracts

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Soy Phospholipid Mixture comprises phosphatidylcholine, phosphatidylethanolamine, phosphatidylinositol, phosphatidic acid and lysophosphatidylcholine. Soybean phospholipids are by-products generated during soybean oil processing.[1]

Application

Soy Phospholipid Mixture has been used as a purified lipid standard in thin layer chromatography for the quantification of whole-cell lysates and mitochondrial lipids from yeast cells.[2]

Biochem/physiol Actions

Phosphatidylcholine is an acetylcholine synthesis substrate. It nourishes the brain. Phosphatidylinositol plays a key role in neural transmission by acting as a secondary messenger precursor. Phosphatidylethanolamine participates in membrane fusion events. The biocompatible and amphiphilic functionality of phospholipids enable them to be a perfect drug delivery system. They are used in generation of the liposomes for carrying drugs.[3] Phospholipids possess hydrophilic colloid property.[1]

Packaging

5 mL Clear Glass Sealed Ampule (690050P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soybean phospholipids
Recent trends for enhancing the diversity and quality of soybean products (2011)
CLD1 Reverses the Ubiquinone Insufficiency of Mutant cat5/coq7 in a Saccharomyces cerevisiae Model System
Kar A, et al.
PLoS ONE, 11(9), 81-98 (2016)
A review on phospholipids and their main applications in drug delivery systems
Li J, et al.
Asian Journal of Pharmaceutical Sciences , 10(2), 81-98 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service