Skip to Content
Merck
All Photos(1)

Key Documents

640009

Avanti

siRNA Delivery Kit

Avanti Research - A Croda Brand 640009, powder

Synonym(s):

siRNA Delivery Kit, Delivery System for siRNA/DNA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$913.00

$913.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$913.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

$913.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand 640009

packaging

1 ea of 1 × (with stopper and crimp cap)

shipped in

dry ice

storage temp.

−20°C

General description

The self-assembly of custom-synthesized MVL5 and GMO with siRNA results in the formation of a double-gyroid bicontinuous inverted cubic phase with colocalized lipid/siRNA domains. The high charge density (due to MVL5) and positive Gaussian modulus of the GMO-containing membranes confer optimal electrostatic and elastic properties for endosomal escape, enabling efficient siRNA delivery and effective, specific gene silencing.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (640009-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecília Leal et al.
Langmuir : the ACS journal of surfaces and colloids, 27(12), 7691-7697 (2011-05-27)
The self-assembly of a custom-synthesized pentavalent cationic lipid (MVL5) and glycerol monooleate (GMO) with small interfering RNA (siRNA) results in the formation of a double-gyroid bicontinuous inverted cubic phase with colocalized lipid/siRNA domains as shown by synchrotron X-ray scattering and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service