330734W
Avanti
SphingoSPLASH™ I
Avanti Research™ - A Croda Brand
Synonym(s):
SphingoSPLASH I LIPIDOMIX™ Quantitative Mass Spec Internal Standard
Select a Size
Select a Size
About This Item
Recommended Products
Assay
>99% (TLC)
form
methanol solution
packaging
package of 1 × 1 ea
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C (−15°C to −25°C)
Packaging
Components
Legal Information
Kit Components Also Available Separately
- 860681PC15 Ceramide-d7 (d18:1-d7/15:0), Avanti Research™ - A Croda Brand 860681P, powder 10µMSDS
- 330729PC15 Glucosyl(β) Ceramide-d7 (d18:1-D7/15:0), Avanti Research™ - A Croda Brand 10µMSDS
- 330728PC13 Galactosyl(β) Ceramide-d7 (d18:1-D7/13:0), Avanti Research™ - A Croda Brand 10µMSDS
- 330727PC15 Lactosyl(β) Ceramide-d7 (d18:1-D7/15:0), Avanti Research™ - A Croda Brand 10µMSDS
- 330726PC13-dihydroceramide-d7(d18:0-d7/13:0), Avanti Research™ - A Croda Brand 10µMSDS
- 860686P15:0 SM(d18:1/15:0)-d9, Avanti Research™ - A Croda Brand (860686P), powder 10µMSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service