330708X
Avanti
Differential Ion Mobility System Suitability LIPIDOMIX™ Kit
Synonym(s):
Differential Ion Mobility System Suitability LIPIDOMIX™ Kit
About This Item
Recommended Products
biological source
bovine heart
form
chloroform:methanol solution
manufacturer/tradename
Avanti Research™ - A Croda Brand
application(s)
lipidomics
metabolomics
shipped in
dry ice
storage temp.
−20°C
General description
Application
Components
Bovine Heart Extract is supplied in one vial as 25 mg in 1 mL of chloroform.
The second vial is supplied in 0.5 mL of chloroform:methanol (1:1) and contains the following components each at 1 mg/mL unless otherwise noted.
- Cholesterol
- 18:1 TG
- 18:1 SM
- 18:1 Ceramide
- 18:1 Lyso PC
- 19:0 Chol Ester
- 14:1 DG
- 14:1 CA
- 14:1 PC
- 14:1 PS
- 14:1 PG
- 14:1 PE
- 14:1 PI (0.25 mg/mL)
- 14:1 PA
Legal Information
Kit Components Also Available Separately
- 171201CHeart Extract Total, Avanti Research™ - A Croda BrandSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3
Target Organs
Eyes,Central nervous system, Liver,Kidney, Respiratory system
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service