Skip to Content
Merck
All Photos(1)

Key Documents

330687

Avanti

Cer1P ELISA (Clear)

Avanti Research - A Croda Brand

Synonym(s):

Cer1P ELISA, C16 Ceramide-1-Phosphate ELISA Snoopers, Clear, for use in absorbance applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

form

liquid

manufacturer/tradename

Avanti Research - A Croda Brand

packaging

1 ea of 1 × (ELISA strip packaged in foil pouch)

shipped in

wet ice

storage temp.

2-8°C

General description

Avanti Lipid Snoopers are lipid microarrays useful in semi-quantitative measurements. These snoopers are relatively stable and provide a higher sensitivity compared to the conventional enzyme-linked immunosorbent assays (ELISA) methods.[1] Ceramide-1-Phosphate (Cer1P) is a bioactive anionic sphingolipid.[2] Ceramide kinase (CERK) mediates the synthesis of Cer1P.[3] Cer1P is also synthesized by the hydrolysis of sphingomyelin in the presence of enzyme sphingomyelinase (SMase).[4]
ELISA Snoopers are 8-well strips pre-coated with a single high-quality lipid species. They can be used to investigate protein-lipid interactions, and characterize and titrate lipid binding proteins. This format provides the following advantages:
  • quantitative lipid-protein binding studies
  • assemble a 96-well plate using only the lipids of interest for binding studies
  • flexibility - use 8 to 96 wells, and 1 to 12 lipids as needed.

The 8-well strips are available in optically clear, white, or black surfaces for use in colormetric, luminescent and fluorescent assays, respectively. Avanti Lipid Snoopers ELISA Strips allow multiple sample conditions to be probed simultaneously in a single plate making them a perfect complement to Avanti Lipid Snoopers Nitrocellulose Strips.

Biochem/physiol Actions

Ceramide-1-Phosphate (Cer1P) mediates macrophage migration, regulates autocrine and paracrine mediators.[2] Elevated levels of Cer1P leads to negative regulation of ceramide kinase (CERK). The intracellular calcium levels modulate Cer1P and CERK functionality.[3] Cer1P is also a crucial regulator of phagocytosis, inflammation, apoptosis and mitosis.[5] Avanti Lipid Snoopers have been used successfully in elucidating the role of annexin V in membrane trafficking.[1]

Packaging

ELISA Strip Packaged in Foil Pouch (330687-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Snoopers is a trademark of Avanti Polar Lipids, LLC

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide 1-phosphate mediates endothelial cell invasion via the annexin a2-p11 heterotetrameric protein complex
Hankins JL, et al.
Test, 288(27), 19726-19738 (2013)
Regulation of the amount of ceramide-1-phosphate synthesized in differentiated human podocytes
Mallela SK, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1864(12), 158517-158517 (2019)
A Nonradioactive Fluorimetric SPE-Based Ceramide Kinase Assay Using NBD-C6-Ceramide
Van O, et al.
Journal of Lipids, 2012 (2012)
Ceramide-1-phosphate, a new bioactive sphingolipid in regulating cell signaling
Marcu AC and Chalfant CE
Future Lipidology , 2(1), 75-84 (2007)
Avanti lipid tools: connecting lipids, technology, and cell biology
Sims KH, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1841(8), 1038-1048 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service