181131C
Avanti
Sunflower PG
Avanti Research™ - A Croda Brand
Synonym(s):
1,2-di-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) PG(18:2(9Z,12Z)/18:2(9Z,12Z))
About This Item
Recommended Products
biological source
(Sunflower)
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 25 mg (181131C-25MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
10 mg/mL
density
1.49 g/cm3
functional group
phospholipid
lipid type
phospholipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Na+].CCCCCCCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC(O)CO)OC(=O)CCCCCCC\C=C/C\C=C/CCCCC
InChI key
JBOGSUDWCXWRFD-YBBFZHODSA-M
Application
Packaging
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
Target Organs
Central nervous system, Liver,Kidney
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service