Skip to Content
Merck
All Photos(1)

Documents

APOH93ED5E5E

Apollo Scientific Ltd

5-Bromo-2-(cyclopropyl)thiophene-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
247.11
MDL number:

Iupac Name

5-bromo-2-cyclopropylthiophene-3-carboxylic acid

Inchi Code

1S/C8H7BrO2S/c9-6-3-5(8(10)11)7(12-6)4-1-2-4/h3-4H,1-2H2,(H,10,11)

InChI key

HYSJHIPLWHUKEE-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/5-bromo-2-cyclopropylthiophene-3-carboxylic-acid

Pictograms

Signal Word

TBC

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/5-bromo-2-cyclopropylthiophene-3-carboxylic-acid


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service