Skip to Content
Merck
All Photos(1)

Documents

APOH11A9D423

Apollo Scientific Ltd

4,4'-Diaminodiphenylamine sulphate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
315.35
MDL number:

bp

300

Iupac Name

sulfuric acid compound with N~1~-(4-aminophenyl)-1,4-benzenediamine (1:1) hydrate

Inchi Code

1S/C12H13N3.H2O4S.H2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12;1-5(2,3)4;/h1-8,15H,13-14H2;(H2,1,2,3,4);1H2

InChI key

UYXSZRUMZNDZKD-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/44-diaminodiphenylamine-sulphate-hydrate

Boiling point

300

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/44-diaminodiphenylamine-sulphate-hydrate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service