Skip to Content
Merck
All Photos(1)

Documents

APO465886928

Apollo Scientific Ltd

8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
282.61
MDL number:

Iupac Name

8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid hydrate

Inchi Code

1S/C9H4ClF3N2O2.H2O/c10-5-1-4(9(11,12)13)2-15-3-6(8(16)17)14-7(5)15;/h1-3H,(H,16,17);1H2

InChI key

HBDANCBTRVDEBT-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethylimidazo12-apyridine-2-carboxylic-acid-hydrate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/8-chloro-6-trifluoromethylimidazo12-apyridine-2-carboxylic-acid-hydrate


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service