Key Documents
Z763292
Sigma-Aldrich
Aldrich® Sure/Seal septa, EPDM rubber with PTFE face
pkg of 10 ea
Synonym(s):
Sure/seal bottles, Sure/seal septa
About This Item
UNSPSC Code:
41121800
packaging
pkg of 10 ea
General description
Dimensions: 26.2 mm diameter x 5 mm thick at center x 1.4 mm thick at rim.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)