Documents
Z759295
Sigma-Aldrich
Joint clip, standard taper
joint: 14, yellow polyethylene
Synonym(s):
NS joint clamps, NS joint clips, National Standard joint clamps, National standard joint clips
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
material
yellow polyethylene
Joint
joint: 14
packaging
pkg of 50 ea
General description
Color coded, rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode; withstands temperatures up to 120 °C.
Visit the Document Library
Customers Also Viewed
Slide 1 of 1