Skip to Content
Merck
All Photos(1)

Key Documents

Z755648

Flat head for Mortexer

pkg of 1 ea, diam. 3 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$55.30

$55.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$55.30

About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

$55.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Benchmark Scientific BV1000-FLAT

diam.

3 in.

General description

Optional retrofit 3 inch Flat head for Mortexer vortexer.

Legal Information

Mortexer is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service