The Aldrich Kugelrohr glassware set, product Z748293, is a 25 mL set-up consisting of a load flask, receiver flask and straight tube. It has an assembled length of 490 mm, and approximately 195 mm extends from the side of the oven.The 100 mL set-up has an assembled length of 515 mm approximately, and 205 mm extends from the side of the oven.
Z748293
Aldrich® Kugelrohr glassware set
25 mL and 100 mL load and ball-tube flasks with straight tube, joint: ST/NS 14/20
Synonym(s):
distillation apparatus, distilling apparatus, fractional distillation apparatus, glass distillation apparatus, kugelrohr apparatus, kugelrohr distillation, short path distillation apparatus, vacuum distillation apparatus
Select a Size
$498.00
Select a Size
About This Item
$498.00
Recommended Products
Joint
joint: ST/NS14/20
packaging
set of 1 pkg
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
-
What are the dimensions of Z748293, Aldrich® Kugelrohr glassware set?
1 answer-
Helpful?
-